missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Respiratory Protection, N5500 Elastomeric Halfmask, Class 1

Product Code. 15816845
Click to view available options
Size:
Large
Medium
Small
Unit Size:
Each
Pack of 12
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15816845

Brand: Honeywell™ N65550031

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-Performance, Dual-Cartridge (Class 1) Half-Mask Respirator

  • Lightweight and low-profile ergonomic design encourages high levels of user acceptance
  • Unique cradle head band and yoke piece for ease of adjustment
  • Contoured revert seal ensures an excellent face seal that will not cause discomfort
  • Thermoplastic elastomer material avoids latex allergens that are common with economy masks
  • Single unit, four-point harness with neck strap is easy to use
  • Harness's ergonomic design and improved suspension system fits perfectly on top of the head and offers a high level of comfort
  • Superior exhalation valve allows more breath to be exhaled makes workers feel cooler and more comfortable
  • Reduced dead air space means less re-breathed exhaled air
  • Variable thickness in sealing area conforms to nose bridge
  • Filter cartridges not included, available for order separately

Order Info

Filter cartridges not included, available for order separately

TRUSTED_SUSTAINABILITY

Specifications

Cartridge/Filter Filter Cartridges Sold Separately
Cartridge Type Dual Cartridge: Class 1
Certifications/Compliance EC Category PPE: 3, EC Certificate: 251, EC Attestation: CE Ceert 251-5500Respir-Dec 2007, EN 140, Declaration of conformity INSPEC N65550031
Color Gray
Material Thermoplastic Elastomer
Size Small
For Use With (Application) Chemical, Laboratory, and Petrochemical Industries
Includes 4-Point Head Harness
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt