Learn More
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas
The A1B1E1K1 cartridge is part of the wide choice of cartridges offered to protect against many hazardous gases, vapors in many applications. The range provides high quality and a cost-effective protection.
Brand: Honeywell™ N06575009L
Description
- Protects against organic vapor, acid gas, ammonia
- Excellent filtration efficiency over extensive period of time
- Lower profile and lightweight for optimal comfort
- Low profile does not obstruct the field of vision
- Threaded connector is easy to attach
Specifications
Class 1 plastic cartridge | |
Protects against organic vapor, acid gas, ammonia | |
99.6 g |
EC Category PPE: 3, QA: ISO 9001/2000, EC Certification: 2259, EC Attestation: 2259, EN 2259 | |
6 Pair/Box, 8 Boxes/Cs. |
Product Content Correction
Product Title
Honeywell™ Class 1 Plastic Cartridge A1B1E1 for Organic Vapors and Acid Gas > Protects against organic vapor, acid gas, ammonia
Spot an opportunity for improvement?Share a Content Correction