Learn More
Honeywell™ Respiratory Protection, N5500 Elastomeric Halfmask, Class 1
High-Performance, Dual-Cartridge (Class 1) Half-Mask Respirator
Brand: Honeywell™ N65550032
This item is not returnable.
View return policy
Description
- Lightweight and low-profile ergonomic design encourages high levels of user acceptance
- Unique cradle head band and yoke piece for ease of adjustment
- Contoured revert seal ensures an excellent face seal that will not cause discomfort
- Thermoplastic elastomer material avoids latex allergens that are common with economy masks
- Single unit, four-point harness with neck strap is easy to use
- Harness's ergonomic design and improved suspension system fits perfectly on top of the head and offers a high level of comfort
- Superior exhalation valve allows more breath to be exhaled makes workers feel cooler and more comfortable
- Reduced dead air space means less re-breathed exhaled air
- Variable thickness in sealing area conforms to nose bridge
- Filter cartridges not included, available for order separately
Order Info
Filter cartridges not included, available for order separately
Specifications
Filter Cartridges Sold Separately | |
EC Category PPE: 3, EC Certificate: 251, EC Attestation: CE Ceert 251-5500 Respir-Dec 2007, EN 140, Declaration of conformity INSPEC N65550031 | |
Thermoplastic Elastomer | |
Chemical, Laboratory, and Petrochemical Industries |
Dual Cartridge: Class 1 | |
Gray | |
Medium | |
4-Point Head Harness |