Learn More
BINDER™ Class 3.1 Independent Temperature Safety Device
Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
Brand:  BINDER™ 8012-1643
 View more versions of this product
Product Code. 16454349
  • £545.00 / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class 3.1
  • With visual alarm
  • Safety devices also available in Class 2 and 3.3
Specifications
Specifications
Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
BINDER Chambers FD 56, FD 115, FD 260, BINDER Chambers FD 720
Temperature Calibration Certificate
Temperature Regulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Class 3.1 Independent Temperature Safety Device > For Use With: FD 56, FD 115, FD 260, FD 720