Learn More
BINDER™ Class 3.1 Independent Temperature Safety Device
Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
Brand: BINDER™ 8012-1644
This item is not returnable.
View return policy
Description
- Class 3.1
- With visual alarm
- Safety devices also available in Class 2 and 3.3
Specifications
Class 3.1 independent temperature safety device - with visual alarm (DIN 12880) | |
BINDER Chambers FED 56, FED 115, FED 260, FED 720 | |
Temperature Calibration Certificate | |
Temperature Regulation |
Product Content Correction
Product Title
BINDER™ Class 3.1 Independent Temperature Safety Device > For Use With: FED 56, FED 115, FED 260, FED 720
Spot an opportunity for improvement?Share a Content Correction