Learn More
Manutan Class 5.2 organic peroxide IATA/ICAO labels, 100mm x 100mm
Brand: Manutan A352646
Code : Z3
Additional Details : Weight : 1.00000kg
Description
A range of unique labels with varying class numbers. Universally agreed airline standards to identify and safely transport dangerous goods by air. Used and recognised by all major carriers and forwarders. Printed on self-adhesive vinyl tape, with clear polypropylene over-laminate.- Organic peroxide
- Self-adhesive vinyl
- Dimensions (L x H) : 100mm x 100mm
- Class 5.2