Learn More
Hirschmann™ Class A Measuring Cylinders, Blue Graduation
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation are manufactured from durable DURAN™ glass tubing.
£26.00 - £179.57
Specifications
Base Type | Hexagonal |
---|---|
Certifications/Compliance | DIN EN ISO 4788 |
Class | A |
Type | Graduated Measuring Cylinder |
Scale Color | Blue |
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
11786482
|
Hirschmann™
2240154 |
5 mL | Certified Conformity |
£41.25
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11748041
|
Hirschmann™
2240160 |
10 mL | Certified Conformity |
£42.05
Pack of 2 |
In Stock
Log in to see stock. |
||||
11758041
|
Hirschmann™
2240170 |
25 mL | Certified Conformity |
£35.80
Pack of 2 |
In Stock
Log in to see stock. |
||||
11768041
|
Hirschmann™
2240175 |
50 mL | Certified Conformity |
£37.25
Pack of 2 |
In Stock
Log in to see stock. |
||||
11778041
|
Hirschmann™
2240180 |
100 mL | Certified Conformity |
£42.26
Pack of 2 |
In Stock
Log in to see stock. |
||||
11788041
|
Hirschmann™
2240186 |
250 mL | Certified Conformity |
£76.45
Pack of 2 |
In Stock
Log in to see stock. |
||||
11798041
|
Hirschmann™
2240190 |
500 mL | Certified Conformity |
£133.00
Pack of 2 |
In Stock
Log in to see stock. |
||||
10569242
|
Hirschmann™
2240193 |
1000 mL | Certified Conformity |
£93.45
Each |
In Stock
Log in to see stock. |
||||
10440261
|
Hirschmann™
2240194 |
2000 mL | Certified Conformity |
£179.57
Each |
In Stock
Log in to see stock. |
||||
10747793
|
Hirschmann™
224017027 |
25 mL | USP |
£26.00
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10097262
|
Hirschmann™
224017527 |
50 mL | USP |
£30.50
Pack of 2 |
In Stock
Log in to see stock. |
||||
10597193
|
Hirschmann™
224018027 |
100 mL | USP |
£39.50
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10765683
|
Hirschmann™
224018627 |
250 mL | USP |
£60.50
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10436973
|
Hirschmann™
224019027 |
500 mL | USP |
£101.00
Pack of 2 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||||
10241253
|
Hirschmann™
224019327 |
1000 mL | USP |
£77.95
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
10332933
|
Hirschmann™
224019427 |
2000 mL | USP |
£147.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
Description
- With spout, main point ring graduation
- Blue graduation
- Conformity certified, with dated batch identification
- Acc. to DIN EN ISO 4788
- Corrosion-free and resistant to thermal shock
Specifications
Hexagonal | |
A | |
Blue | |
Glass |
DIN EN ISO 4788 | |
Graduated Measuring Cylinder | |
Round |