Learn More
Hirschmann™ Class AS Volumetric Pipettes with Amber Stain Graduation
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Volumetric Pipettes with Amber Stain Graduation are manufactured from clear AR™ glass.
£47.45 - £161.00
Specifications
Class | AS |
---|---|
Material | AR-GLAS |
Graduated | Yes |
Graduation Color | Brown |
Type | Volumetric Pipet |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
11782252
|
Hirschmann™
1340129 |
0.5 mL |
£114.00
Pack of 12 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
||||||
11712262
|
Hirschmann™
1340180 |
100 mL |
£161.00
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11834602
|
Hirschmann™
1340139 |
1.5 mL |
£160.00
Pack of 12 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
11708001
|
Hirschmann™
1340160 |
10 mL |
£74.45
Pack of 6 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
||||||
11702262
|
Hirschmann™
1340164 |
15 mL |
£85.90
Pack of 6 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
||||||
11727981
|
Hirschmann™
1340133 |
1 mL |
£109.00
Pack of 12 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||||
11786652
|
Hirschmann™
1340145 |
2.5 mL |
£52.26
Pack of 6 |
Estimated Shipment: 16-04-2025
Log in to see stock. |
||||||
11718001
|
Hirschmann™
1340165 |
20 mL |
£90.60
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11728001
|
Hirschmann™
1340170 |
25 mL |
£92.35
Pack of 6 |
In Stock
Log in to see stock. |
||||||
12771158
|
Hirschmann™
1340148 |
3 mL |
£47.45
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11706662
|
Hirschmann™
1340173 |
30 mL |
£109.00
Pack of 6 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
||||||
11736672
|
Hirschmann™
1340174 |
40 mL |
£141.00
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11796652
|
Hirschmann™
1340150 |
4 mL |
£55.80
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11738001
|
Hirschmann™
1340175 |
50 mL |
£141.00
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11797991
|
Hirschmann™
1340155 |
5 mL |
£63.70
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11823141
|
Hirschmann™
1340159 |
9 mL |
£84.00
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11716133
|
Hirschmann™
1340156 |
6 mL |
£85.95
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11787991
|
Hirschmann™
1340140 |
2 mL |
£89.35
Pack of 12 |
In Stock
Log in to see stock. |
||||||
10448201
|
Hirschmann™
1340157 |
7 mL |
£55.05
Pack of 6 |
In Stock
Log in to see stock. |
||||||
11726133
|
Hirschmann™
1340158 |
8 mL |
£74.85
Pack of 6 |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
Description
- One mark
- Acc. to DIN EN ISO 648
- Conformity certified, with dated batch identification
Specifications
AS | |
Yes | |
Volumetric Pipet |
AR-GLAS | |
Brown |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hirschmann™ Class AS Volumetric Pipettes with Amber Stain Graduation