Learn More
Kartell™ Class B (TPX™) PMP Low Form Beakers
Accurately measures the volume of a liquid. Kartell™ Low Form Graduated Beakers Class B uses horizontal marked lines to assist in measuring out various volumes of a liquid.
Brand:  Kartell™ 0154200
Product Code. 10325731
  • ONLINE EXCLUSIVE PRICE
    £81.55
    £80.07 / Pack of 20
    Save £1.48
    1% Off
Only null left
Estimated Shipment: 22-01-2026
Add to basket
Description
Description
Crystal clear, will withstand 170°C for short periods, 150°C for continuous work. Permanent graduations molded in for reproducibility, excellent chemical resistance. Not suitable for use with stirrer hotplates. Autoclavable.