missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class E2 Weight with COFRAC Calibration Certificate

Calibrate weights of lower accuracy classes. Zwiebel™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.

Brand:  Zwiebel™ PBI2302P

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11780633

  • £220.48 / Each
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

  • Class E2 calibration weight
  • In compliance with OIML R111
  • Delivered with a COFRAC calibration certificate guaranteeing traceability
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications

Specifications

20 mg
±0.01 mg
For use with Weighing Instruments of Special Accuracy Class I
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers