missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class F1 Box Weights with Certificate

Use with weighing instruments of special accuracy class I and high accuracy class II. Zwiebel™ Class F1 Weights are intended for use in the verification or calibration of class F2 weights.

Brand:  Zwiebel™ PBI23135

 View more versions of this product

Product Code. 11750793

  • £202.46 / Each
Estimated Shipment: 04-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class F1 calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifications

Specifications

100 g
±0.50 mg
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers