missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class F1 Box Weights with Certificate

Use with weighing instruments of special accuracy class I and high accuracy class II. Zwiebel™ Class F1 Weights are intended for use in the verification or calibration of class F2 weights.

Brand:  Zwiebel™ PBI23129

 View more versions of this product

Product Code. 11760773

  • £152.64 / Each
Estimated Shipment: 08-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class F1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications

    Specifications

    ±0.1 mg
    For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    View accessory products
    Special Offers

    Special Offers