missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class M1 Box Weights
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Box Weights are ideal for the verification or calibration of class M₂ weights.
£19.20 - £1120.00
Specifications
For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |
---|
Product Code | Brand | Tolerance | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tolerance | Price | Quantity & Availability | ||||||
11770743
|
Zwiebel™
PBI23100 |
±50000 mg |
£1120.00
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11740593
|
Zwiebel™
PBI2300H |
±0.5 mg |
£19.20
Each |
In Stock
Log in to see stock. |
||||||
11768204
|
Zwiebel™
PBI2811M |
±2 mg |
£26.25
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11710723
|
Zwiebel™
PBI23094 |
±5 mg |
£42.45
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
11748204
|
Zwiebel™
PBI2811K |
±1.2 mg |
£23.25
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
11720723
|
Zwiebel™
PBI23095 |
±10 mg |
£54.55
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
||||||
11760723
|
Zwiebel™
PBI23099 |
±250 mg |
£648.00
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11758204
|
Zwiebel™
PBI2811L |
±1.6 mg |
£24.25
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11700593
|
Zwiebel™
PBI2300C |
±0.2 mg |
£19.20
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
Description
- Class M1 weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zwiebel™ Class M1 Box Weights