missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M1 Box Weights with Certificate

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Box Weights are ideal for the verification or calibration of class M₂ weights.

Brand:  Zwiebel™ PBI23116

 View more versions of this product

Product Code. 11730763

  • £95.95 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications

    Specifications

    ±1 mg
    For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    View accessory products
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.