missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class M1 Weights with Certificate
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.
£87.85 - £839.00
Specifications
For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |
---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
11730603
|
Zwiebel™
PBI2300W |
100 mg | ±0.5 mg |
£87.85
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
11774744
|
Zwiebel™
PBI23723 |
10 kg | ±500 mg |
£264.00
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
||||
11784744
|
Zwiebel™
PBI23724 |
20 kg | ±1000 mg |
£314.00
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
11770593
|
Zwiebel™
PBI2300M |
1 mg | ±0.2 mg |
£88.85
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
11740603
|
Zwiebel™
PBI2300X |
200 mg | ±0.6 mg |
£88.85
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
11700603
|
Zwiebel™
PBI2300R |
10 mg | ±0.25 mg |
£88.85
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||||
11794744
|
Zwiebel™
PBI23726 |
50 kg | ±2500 mg |
£839.00
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
11764744
|
Zwiebel™
PBI23721 |
5 kg | ±250 mg |
£210.00
Each |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||||
Description
- Class M1 calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |