Learn More
Pangea™ Class Rock Pack
Brand: Pangea™
Code : 07
Additional Details : Weight : 2.00000kg
Specifications
Class Rock Pack | |
Limestone; sandstone; slate; porphyry; basalt; granite; chalk; gabbro; marble; tub of clay soil; tub of sandy soil | |
1/Ea. | |
For Identifying Rocks, Including Separation the basis of Grain Size, Textures, Structures, Color, Mouldability and Calcite and Silica Content |