Learn More
Emma Safety Footwear Clay S3 Shoe
If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay.
Brand:  Emma Safety Footwear MM98156946
 View more versions of this product
Product Code. 17502646
  • £114.00 / 1 pair
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay. A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel.
  • This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications
Specifications
ISO 20345:2011
Black
Hydro-Tec Antibacterial
46
Low-cut
Laces
Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Nubuck, Oiled Nubuck
Extra-wide Width
Safety Shoe