missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Clay S3 Shoe

Product Code. 17552636
Click to view available options
Size:
39
40
41
42
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17552636

Brand: Emma Safety Footwear MM98054948

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay.

  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay. A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel.
  • This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
Material Nubuck, Oiled Nubuck
Size 48
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.