Learn More
Vici Valco Instruments™ Clean-Cut™ LC Tubing Cutter
It is designed to cut PTFE, ETFE and polymers in general and PEEK capillary tubing in particular.
Brand:  Vici Valco Instruments™ JR-797
Product Code. 12762680
  • £85.95 / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cuts PEEK, PTFE, ETFE, and another polymeric tubing
  • No distortion to OD or ID
  • Ideal for the most demanding LC applications
  • Safety lock secures the blade
  • Right angles and clean cuts are essential in LC to avoid leaks and dead volume.
  • This handy tool makes it easy to get burr-free, perpendicular ends on polymeric tubing without distorting the outside diameter and/or closing the inside diameter of the tubing.
  • The compact design and unique safety locking mechanism allow every chromatographer to keep one in their pocket.
Specifications
Specifications
Clean-cut Tubing Cutter
For 1/16 in., 1/8 in., 2 mm, 3 mm, and 4 mm OD tubing