Learn More
Abeba™ Clean Room 3620 Shoes
Special ESD clean room shoes.
Brand: Abeba™ 3620/36
Additional Details : Weight : 0.94000kg
Description
- CE EN ISO 20347:2012 OB A, E SRA
- Microfiber
- White
- ESD dissipative according to DIN EN 61340
- Fixed gaitor, cleanroom textile
Specifications
CE EN ISO 20347:2012 OB A, E SRA | |
Microfiber | |
3620 |
White | |
36 |