Learn More
Abeba™ Clean Room 3620 Shoes
Special ESD clean room shoes.
Brand:  Abeba™ 3620/46
Additional Details : Weight : 0.94000kg
 View more versions of this product
Product Code. 15610388
  • £81.31 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRA
  • Microfiber
  • White
  • ESD dissipative according to DIN EN 61340
  • Fixed gaitor, cleanroom textile
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRA
Microfiber
3620
White
46