Learn More
Sampling Systems™ Cleaning Coupons for Metal
Test and validate your cleaning regime with this range of cleaning validation coupons for various metals.
Brand:  Sampling Systems™ 8644S6-100S
 View more versions of this product
Product Code. 15829561
  • £144.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tests available for aluminum, 304 stainless steel and 316 stainless steel
  • All metals also available for either bright polish or satin finish
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
Cleaning Coupon
Satin finish
Stainless Steel (316)
100 x 100 mm
Testing and validating cleaning regime