missing translation for 'onlineSavingsMsg'
Learn More

BCR™ Cleanroom Face Masks

Product Code. p-7227757 Shop All Berkshire Products
Click to view available options
Dimensions (L x W):
180 x 90 mm
210 x 90 mm
Size:
Medium
Large
Packaging Type:
Bulk Packaged
Individually Bagged
Fastening Type:
Earloops
Headband
Unit Size:
Pack of 1050
Pack of 1500
Pack of 1800
This item is not returnable. View return policy

Product Code. 17871690

Brand: BCR™ BM180ELK25I

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Berkshire masks provide filtration efficiency while allowing long wearing comfort for operators, and are designed to protect your cleanroom or controlled environment from human contamination

  • Extra wide body of mask reduces exposed skin and provides better coverage for larger faces
  • Filter Medium: Meltblown polypropylene provides superior particle and bacterial filtration efficiency
  • Ultrasonic bonding reduces contamination and outgassing

Specifications

Product Type Cleanroom Face Mask
Color Light Blue
Fastening Type Earloops
Material Polypropylene
Size Medium
Sterility Non-sterile
Dimensions (L x W) 180 x 90 mm
Certifications/Compliance ASTM F2299, ASTM F2101
Cleanroom Class Class 10-100,000 (ISO 4-8)
For Use With (Application) Industrial Manufacturing, Medical Device, Microelectronics, Pharmacy, Food Processing
Model BM180ELK
Packaging Type Individually Bagged
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.