Learn More
CleanSpace™ CleanSpace2™ Power System
Battery-powered, positive pressure air-purifying respirator (PAPR). The CleanSpace2 respirator can be used with CleanSpace Half Masks (PAF-0033, PAF-1010, PAF-0027). Compatible with the full range of standard CleanSpace particulate (P3) and combined gas filters and pre-filters.
Brand:  CleanSpace™ PAF-0034
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15910102
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Light weight and comfortable
  • AirSensit™ System for mask pressure control and breath responsive airflow
  • One-button system and standby and auto-start/stop mode for quick don/doffing
  • Easy to use support harness
  • Altitude compensated filter blocked detection
  • Long operating time: up to 8 hours and quick battery recharging: < 2hours
  • Can be tested by Quantitatively Mask Fit (PortaCount) Tests
  • Compatible with all CleanSpace Half Masks
  • Compatible with all CleanSpace filters: Particulate and Combined Gas Filters
Specifications
Specifications
PAPR Assembly, No Headtop
PAF-0034
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen
Mask sold separately