Learn More
CleanSpace Technology™ CleanSpace3™ Cleaning and Storage Plug Set
Tool for use with the CleanSpace™ HALO Respirator unit (CS3000). Seals all the openings on the CleanSpace™ HALO PAPR unit for storage. Made of polycarbonate and silicone.
Brand: CleanSpace Technology™ CS3011
35.34 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Additional Details : Weight : 0.06300kg
Alert:
Do not wear the respirator when the plug set is in place.
Description
- Used with the revolutionary CleanSpace HALO PAPR: light weight, no hoses or belts
- Designed for comfort over long periods
- Allows sweating and breathability
- Easy to wash and quick drying
- Designed for long wear in harsh environments
- Easy and quick replacement
Specifications
CS3011 | |
CleanSpace™ HALO Respirator unit (CS3000) |