Learn More
CleanSpace™ Technology CleanSpace3™ Head Harness for Half Mask (non fabric)
Replacement part for use with the CleanSpace HALO. Consists of TPU clips, polyethylene strap with polyester adjustment band.
Brand: CleanSpace™ Technology CS3008
Description
- Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
- Designed for comfort over long periods
- Allows sweating and breathability
- Multiple adjustment points
- Easy to wash and quick drying
- Durable and designed for long wear in harsh environments
- Easy and quick replacement
Specifications
Reusable | |
Head Harness | |
CS3000 PAPR System | |
CS3008 |