missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates
BIOPHARMA

Product Code. 10737931
Click to view available options
Surface Treatment:
MaxiSorp
PolySorp
Certified:
No
Yes
Unit Size:
Pack of 60
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Product Code. 10737931

Brand: Thermo Scientific™ undefined

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Perform quantitative and qualitative solid phase immunoassays with these nonsterile 96-well plates, available in a variety of binding surfaces.

Perform quantitative and qualitative solid phase immunoassays with Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates, which include Nunc™, Immulon™ and Microtiter™ plates. Choose from a variety of binding surfaces for reliable and reproducible results in colormetric assays.

96-Well Solid Plates

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces

  • PolySorp for adsorption of hydrophobic molecules
  • MaxiSorp is hydrophilic and ideal for antibody sandwich assays

Ordering Information:

Please note that availability may vary by country. 446612 includes certificate.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Hydrophobic
Certified No
Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 60 Cases
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 250 μL
Sterility Non-sterile
Surface Treatment PolySorp
Well Shape Round, C Bottom
No. per Case 60
No. per Pack 5
Plate Blocking None
Target Molecule Lipids
Well Design C96
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.