missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Clear Drying Chambers for FreeZone™ Freeze Dryers

Clear chamber provides visibility for bulk freeze drying.

£2875.00 - £6915.00

Specifications

Color Clear
For Use With (Equipment) FreeZone™ Freeze Dryers
View More Specs

Products 4
Product Code Brand Diameter (English) Weight (English) Height (English) Price Quantity & Availability  
Product Code Brand Diameter (English) Weight (English) Height (English) Price Quantity & Availability  
15721008
View Documents
Labconco™
7318802
17.4 in. 19 lb. 15.3 in.
£6915.00
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
15787807
View Documents
Labconco™
7318700
13.6 in. 17 lb. 24.6 in.
£2875.00
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
15797807
View Documents
Labconco™
7318702
13.6 in. 14 lb. 16.8 in.
£4285.00
Each
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
 
15796968
View Documents Promotion Details
Labconco™
7318800
17.4 in. 22 lb. 23 in.
£5545.00
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Clear chamber provides visibility for bulk freeze drying.

  • 12.0 in. dia. acrylic chamber fits all FreeZone™ Freeze Dryers
  • Provides visibility for bulk freeze drying
  • Accommodates Product Shelves (5 shelves) 7317200
  • Includes clear arylic lid with gasket, vacuum release valve & pass-through for electrical cords
  • Will fit 10.12 in. or 14.0 in. diameter condensers
  • 8 in. diameter opening to collector on bottom of chamber
Specifications

Specifications

Clear
FreeZone™ Freeze Dryers
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.