Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
£145.00 - £1220.00
Specifications
Format | Standard |
---|---|
Detection Method | Colorimetric |
Array | 8 x 12 |
Color | Clear |
Product Type | Microplate |
Alert:
Each lot is functionally tested for IgG binding.
Product Code | Brand | Surface Treatment | Certified | Volume (Metric) Working | Barcode | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Certified | Volume (Metric) Working | Barcode | Price | Quantity & Availability | ||
10547915
|
Thermo Scientific™
3355 |
Immulon 1 B | No | 230 μL | No Barcode |
£151.00
Pack of 50 |
In Stock
Log in to see stock. |
||
10795026
|
Thermo Scientific™
3455 |
Immulon 2 HB | No | 280 μL | No Barcode |
£348.00
Pack of 50 |
In Stock
Log in to see stock. |
||
10468075
|
Thermo Scientific™
3855 |
Immulon 4 HBX | No | 280 μL | No Barcode |
£357.00
Pack of 50 |
In Stock
Log in to see stock. |
||
10394751
|
Thermo Scientific™
439454 |
MaxiSorp | Yes | 350 μL | No Barcode |
£428.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10547781
|
Thermo Scientific™
442404 |
MaxiSorp | No | 350 μL | No Barcode |
£306.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10498802
|
Thermo Scientific™
456529 |
PolySorp | No | 350 μL | No Barcode |
£545.00
Pack of 180 |
In Stock
Log in to see stock. |
||
10294401
|
Thermo Scientific™
456537 |
MaxiSorp | Yes | 350 μL | No Barcode |
£1220.00
Pack of 180 |
In Stock
Log in to see stock. |
||
10264451
|
Thermo Scientific™
460984 |
MaxiSorp | Yes | 350 μL | Includes Barcode |
£644.00
Pack of 180 |
In Stock
Log in to see stock. |
||
10275411
|
Thermo Scientific™
467320 |
MediSorp | Yes | 350 μL | No Barcode |
£446.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10565131
|
Thermo Scientific™
467340 |
MultiSorp | No | 350 μL | No Barcode |
£283.00
Pack of 60 |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||
10449672
|
Thermo Scientific™
475094 |
PolySorp | No | 350 μL | No Barcode |
£350.00
Pack of 60 |
In Stock
Log in to see stock. |
||
10190825
|
Thermo Scientific™
9502227 |
Universal Binding | No | 330 μL | No Barcode |
£145.00
Pack of 50 |
In Stock
Log in to see stock. |
||
10130665
|
Thermo Scientific™
95029330 |
Enhanced Binding | Yes | 330 μL | No Barcode |
£282.00
Pack of 50 |
In Stock
Log in to see stock. |
||
Description
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
- Clear polystyrene
- Choice of four surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
- Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
- Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
Specifications
Standard | |
8 x 12 | |
Microplate | |
96 | |
None |
Colorimetric | |
Clear | |
Polystyrene | |
Non-sterile |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.