Learn More
Hirschmann™ Clear Glass Graduated Pipettes with Amber Stain Graduation
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Clear Glass Graduated Pipettes with Amber Stain Graduation come with a cotton plug end as standard.
Brand:  Hirschmann™ 1000142
 View more versions of this product
Product Code. 11746632
  • £55.10 / Pack of 12
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Short line
  • Acc. to DIN EN ISO 835
  • Graduated to tip, zero on top, type 3
Specifications
Specifications
B
Yes
0.1 mL
12 Pack
360 ±5 mm
Soda Lime Glass
Amber
Amber graduations permanently with silver diffusion color
2 mL
Graduated Pipet