Learn More
Berlin Packaging™ Clear Laboratory Glassware Pot
Product Code. p-8000014
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Quantity:
18 Set
20 Set
48 Set
63 Set
Unit Size:
Pack of 2688
Pack of 4158
Pack of 720
Product Code. 30408780
Brand: Berlin Packaging™ 1000732
Use the Berlin Packaging™ Clear Laboratory Glassware Pot as a fundamental piece of laboratory glassware. The autoclavable clear glass is breakproof and chemically and thermally shockproof.
  • Clear borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Specifications
Certifications/Compliance DIN 68
Quantity 18 Set
Capacity (Metric) 1000 mL
Material Glass
Product Type Pot