Learn More
Corning™ Clear Polystyrene 96-Well EIA/RIA Microplates
GREENER_CHOICE
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
£160.00 - £247.00
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
View More Specs
Products 3
Product Code Brand Well Shape Binding Property Packaging Price Quantity & Availability  
Product Code Brand Well Shape Binding Property Packaging Price Quantity & Availability  
10293532
GREENER_CHOICE
View Documents Promotion Details
Corning™
9018
Flat Bottom (F-well) High Binding 25/Pk.
£247.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
10337891
GREENER_CHOICE
View Documents Promotion Details
Corning™
3591
Flat Bottom (F-well) Medium Binding Individually Wrapped
£160.00
Pack of 50
In Stock
to see stock.
Edge
Add to basket
10380982
GREENER_CHOICE
View Documents Promotion Details
Corning™
3797
Round Medium Binding 25/Pk.
£238.00
Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Specifications
Standard
8 x 12
Polystyrene
75 to 200 μL
Without Lid
Clear
96
Non-sterile
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Corning™ Clear Polystyrene 96-Well EIA/RIA Microplates