missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Clear Rain Gauge

Measure accumulated precipitation with these clear buterate cylindrical rain gauges. Easy to read rain level in English or Metric units

Brand:  Cole-Parmer™ 03319-10

Product Code. 11744819

  • £77.40 / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Cylindrical rain gauge
  • Easy to read
  • English or metric units
  • Meausres up to 9.92 in. rain fall
  • Includes buterate mounting bracket, screws and data recording sheet
  • Recording sheet logs rain fall data for up to one year
Specifications

Specifications

±0.2 mm
14 in.
Measuring rain water
Buterate
4 in.
Buterate mounting bracket, screws, data recording sheet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.