Learn More
Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates
Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.
£130.00 - £186.00
Specifications
Barcode | No Barcode |
---|---|
Format | Standard |
Detection Method | Colorimetric |
Array | 8 x 12 |
Color | Clear |
Alert:
Each lot is functionally tested for IgG binding.
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Price | Quantity & Availability | |||||
10635577
|
Thermo Scientific™
3555 |
Immulon 1 B |
£146.00
Pack of 50 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
10111005
|
Thermo Scientific™
3655 |
Immulon 2 HB |
£183.00
Pack of 50 |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
10450343
|
Thermo Scientific™
449824 |
MaxiSorp |
£186.00
Pack of 60 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
10479382
|
Thermo Scientific™
475434 |
PolySorp |
£130.00
Pack of 60 |
In Stock
Log in to see stock. |
|||||
Description
Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
- Clear polystyrene
- Choice of a variety of surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific PolySorp and Immulon 1B are for adsorption of hydrophobic molecules.
- Thermo Scientific Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
Ordering Information:
Please note that availability may vary by country.
Specifications
No Barcode | |
Colorimetric | |
Clear | |
Polystyrene | |
Non-sterile | |
None |
Standard | |
8 x 12 | |
Microplate | |
96 | |
Round |