Learn More
Fisherbrand™ Clear Straight Sided Glass Jars without Caps
GREENER_CHOICE
Clear, USP Type III soda-lime glass wide mouth design is great for large solid samples and allow for complete removal of contents. Ideal for soil sampling and environmental applications
£18.20 - £381.00
Specifications
Color Clear
Closure Included No
Mouth Wide
View More Specs
Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
16184647
GREENER_CHOICE
View Documents
Fisherbrand™
FB02912304
30 mL
£381.00
Pack of 384
Estimated Shipment: 04-06-2025
to see stock.
Add to basket
16174557
GREENER_CHOICE
View Documents
Fisherbrand™
FB02911996
60 mL
£144.00
Pack of 144
Estimated Shipment: 30-07-2025
to see stock.
Add to basket
16184557
GREENER_CHOICE
View Documents
Fisherbrand™
FB02911997
120 mL
£21.48
Pack of 24
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
16184487
GREENER_CHOICE
View Documents
Fisherbrand™
FB02911876
240 mL
£18.20
Pack of 12
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
16124627
GREENER_CHOICE
View Documents
Fisherbrand™
FB02912118
480 mL
£27.30
Pack of 12
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
16194647
GREENER_CHOICE
View Documents
Fisherbrand™
FB02912305
950 mL
£24.25
Pack of 12
Estimated Shipment: 30-07-2025
to see stock.
Add to basket
Description
Description
  • Straight sided walls allow for the complete removal of contents
  • Available in a wide variety of sizes, from 1 to 32 oz.
Specifications
Specifications
Clear
No
Wide
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Clear Straight Sided Glass Jars without Caps