missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens Flow Detergent

Removes a wide range of organic soils and prevents scale buildup. Diversey ClearKlens Flow Detergent is a low foam, heavy duty CIP caustic detergent.

Brand:  Diversey™ 7513367

Code : NEW

Additional Details : CAS Number : 1310-73-2 Weight : 29.80000kg Transport : UN number : 1824 Chem class : 8 Pack group : II


Product Code. 15300318

  • £141.00 / 20 L
Estimated Shipment: 22-08-2024
to see stock.

Description

Description

  • Heavy duty, hard water tolerant, low foam, formulated liquid caustic detergent for CIP cleaning
  • Used at 0/7 to 7% w/w at temperatures ranging from ambient to 80°C, depending upon application, product residue, and water hardness
  • Specifically formulated to provide scale inhibition protection during the final rinse step
Specifications

Specifications

Liquid
Non-sterile
Unfiltered
>12
Canister
20 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Clearklens Flow VH20

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers