missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ ClearKlens IPA Disinfectant
Disinfects work surfaces and process equipment in cleanrooms. Diversey ClearKlens IPA Disinfectant is sterile and filtered to 0.2μm. The 900mL bottles include a trigger spray.
Brand: Diversey™ 7513400
This item is not returnable.
View return policy
Description
- Ready to use spray for the disinfection of work surfaces and process equipment
- 70% IPA and 30% water for injection
- 900mL bottle sizes come with trigger spray
Specifications
Disinfectant | |
Sterile | |
Filtered | |
6/Cs. |
∼7 | |
70% Isopropyl alcohol, 30% water | |
Trigger spray | |
5.4 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Clearklens IPA VH1
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol