Learn More
Diversey™ ClearKlens IPA VH1 Disinfectant
Ready to use disinfectant for non-porous hard surfaces and gloves
Brand: Diversey™ 101105067
Additional Details : Weight : 5.25000kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
ClearKlens IPA is a non-sterile, ready to use disinfectant for the Life Sciences industry. It is used as general purpose mid-shift and terminal disinfectant. ClearKlens IPA is based on 70% isopropyl alcohol. ClearKlens IPA is available as 500 mL trigger spray, and as 1 L bottle or 5 L canister.
Key properties
- ClearKlens IPA is a fast acting disinfectant spray for use in mid-shift and terminal disinfection applications.
- It has a broad spectrum of efficacy against most vegetative forms of micro- organisms.
- ClearKlens IPA is safe for use on food contact surfaces and evaporates rapidly, leaving no residue. It is suitable for the disinfection of alcohol-safe surfaces, machines and components (conveyors, fillers, bottle inspectors, water-sensitive components, etc.)
- BPR approved pack and formulation
Benefits
- Filtered at 0.2 μm
- Ready to use
- Effective broad-spectrum activity
- Non foaming
- Suitable for use in moisture sensitive areas
- Readily evaporates leaving no residues
Specifications
Surface Disinfectant | |
Non-sterile | |
Spray Bottle | |
12/Cs. |
6 to 9 | |
70% IPA/30% deionized water | |
For AT, BE, CH, DE, FR, GB, IE, IT, NL only | |
500 mL |
Safety and Handling
- ClearKlens IPA VH1
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol