Learn More
Diversey™ ClearKlens NTC Detergent
Provides effective single stage recovery cleaning in hard water with the presence of very high levels of product residue. Diversey ClearKlens NTC Detergent is a heavy duty, caustic, defoamed detergent that can be used in the removal of a wide range of organic and inorganic product residues.
Brand: Diversey™ 7513363
Code : NEW
Additional Details : CAS Number : 1310-73-2 Weight : 25.00000kg Transport : UN number : 1824 Chem class : 8 Pack group : II
Description
- Highly effective at single stage recovery cleaning in hard water with the presence of very high levels of product residue
- Removes a wide range of organic and inorganic product residues
- Hard water tolerant
- Defoamed liquid
Specifications
Liquid | |
Non-sterile | |
Filtered |
>12 | |
Canister | |
20 L |
Safety and Handling
- Clearklens NTC VH18
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide