missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens Spek Detergent

Removes a wide range of organic soils and prevents scale build up in SIP applications, improving cleaning performance. Diversey ClearKlens Spek Detergent is a low foam, caustic detergent. It is hard water tolerant.

Brand:  Diversey™ 7513359

Code : NEW

Additional Details : CAS Number : 1310-73-2 Weight : 29.40000kg Transport : UN number : 1824 Chem class : 8 Pack group : II

Product Code. 15360318

  • £104.00 / 20 L
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

  • Effectively removes a wide range of organic soils
  • Prevents scale build up in CIP applications, improving cleaning performance
  • Used at 0.3 - 5% w/w concentrations
  • Should be rinsed thoroughly after use to remove detergent from product contact surfaces
Specifications

Specifications

Liquid
Non-sterile
20 L
>12
Unfiltered
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers