Learn More
Diversey™ ClearKlens Tego 2000 Disinfectant
Cleans and disinfects in sterile environments. The Diversey ClearKlens Tego 2000 Disinfectant is an active amphoteric disinfectant with a broad spectrum of activity. This microbiocidal disinfectant can be applied by means of a mop or cloth.
Brand: Diversey™ 7516427
259.97 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Description
- Hard surface disinfectant based on amphoretics
- Supplied with certificates of analysis, sterility, and irradiation where applicable
Specifications
∼7 | |
Unfiltered | |
6 x 900 mL |
Sterile | |
SS VH25S | |
900 mL |
Safety and Handling
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.