missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CLED Agar
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
CLED Agar
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
Description
Detect common urinary tract pathogens using Thermo Scientific™ CLED Agar, a traditional media which provides good colonial differentiation, clear diagnostic characteristics and prevents swarming from Proteus spp. due to the electrolyte deficiency in the formulation.
Isolate, enumerate and presumptively identify all UTI related microorganisms with this traditional urinary bacteriology medium which provides a valuable non-inhibitory diagnostic for plate culture.
This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Blue/ Green, clear |
| Content And Storage | 2°C to 10°C |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Media Type | CLED Agar |
| Product Type | Agar |
| Fill Weight | 19.5g ± 1.0g |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction