Learn More
Molecular Probes™ Click-iT™ Plus Alexa Fluor™ 488 Picolyl Azide Toolkit
Click-iT Plus Alexa Fluor 488 Picolyl Azide Toolkit
Brand:  Molecular Probes™ C10641
396.72 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 15403493
  • £522.00 / 1 set
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Click-iT™ Plus Alexa Fluor™ picolyl azide toolkits contain everything you need to perform copper-catalyzed click reactions with compounds that are sensitive to copper, while retaining all of the benefits of the standard azide/alkyne click reaction. Picolyl azides react efficiently with chelated copper, so free copper in the click reaction can be minimized, protecting against undesired copper side reactions with proteins (e.g., GFP, RPE), nucleic acids (e.g., RNA, oligos), and even small molecules (e.g., phalloidin).
  • Flexible—optimize your own Click–iT™ reaction for the detection or labeling of any alkyne-containing biomolecule in vitro or in cells and tissue samples
  • Bright and sensitive—superior fluorescent Alexa Flour™ dyes combined with the efficiency of picolyl azide
  • Convenient—contains all the buffers and reagents needed to create the optimal click reaction
    Each Click-iT™ Plus Picolyl Azide Toolkit includes 100μg of Alexa Fluor™ dye-picolyl azide and all of the reagents required to create a reaction cocktail with the optimal copper and chelate concentration for any click chemistry-based labeling or detection application. Guidelines for designing and optimizing the picolyl azide/alkyne click reaction of your choice are provided in the product manual. Additionally Click-iT™ Plus picolyl azides offer increased sensitivity and faster reaction times compared to standard azides using identical reaction conditions.
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    488 Picolyl Azide Toolkit
    • 1 x 100 μg Alexa Fluor™ dye-picolyl azide
    • 1 x 4 mL Click-iT™ reaction buffer
    • 1 x 1.5 mL copper (II) sulfate
    • 1 x 500 μL copper protectant
    • 1 Click-iT™ buffer additive
    Each k
    Alexa Fluor 488
    1 kit
    Click-iT™
    Alexa Fluor Dyes
    Sugerencias de productos
    Sugerencias de productos
    Videos
    SDS
    Documentos
    Documentos
    Certificados
    Ofertas especiales
    Ofertas especiales
    For Research Use Only. Not for use in diagnostic procedures.