Learn More
Molecular Probes™ Click-iT™ Plus Alexa Fluor™ 555 Picolyl Azide Toolkit
Click-iT Plus Alexa Fluor 555 Picolyl Azide Toolkit
Brand: Molecular Probes™ C10642
Code : NEW
Additional Details : Weight : 0.25000kg
Description
Click-iT™ Plus Alexa Fluor™ picolyl azide toolkits contain everything you need to perform copper-catalyzed click reactions with compounds that are sensitive to copper, while retaining all of the benefits of the standard azide/alkyne click reaction. Picolyl azides react efficiently with chelated copper, so free copper in the click reaction can be minimized, protecting against undesired copper side reactions with proteins (e.g., GFP, RPE), nucleic acids (e.g., RNA, oligos), and even small molecules (e.g., phalloidin).Each Click-iT™ Plus Picolyl Azide Toolkit includes 100μg of Alexa Fluor™ dye-picolyl azide and all of the reagents required to create a reaction cocktail with the optimal copper and chelate concentration for any click chemistry-based labeling or detection application. Guidelines for designing and optimizing the picolyl azide/alkyne click reaction of your choice are provided in the product manual. Additionally Click-iT™ Plus picolyl azides offer increased sensitivity and faster reaction times compared to standard azides using identical reaction conditions.
Specifications
555 Picolyl Azide Toolkit | |
Click-iT™ | |
Alexa Fluor Dyes |
|
|
Alexa Fluor 555 |
For Research Use Only. Not for use in diagnostic procedures.