Learn More
Invitrogen™ Click-iT™ sDIBO Alkynes for copper-free click chemistry
Experience copper-free 'click chemistry' with Click-IT™ Alexa Fluor™, and sDIBO Alkyne reagents, which are ideal for surface labeling of live cells and minimize damage to enzymes and fluorescent proteins.
£298.00 - £576.00
Specifications
Green Features | Less hazardous |
---|---|
Labeling Method | Click Chemistry |
Shipping Condition | Dry Ice |
Labeling Target | Azide |
Product Code | Brand | Label or Dye | Reactive Moiety | Molecular Weight (g/mol) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Label or Dye | Reactive Moiety | Molecular Weight (g/mol) | Price | Quantity & Availability | |||
10774701
|
Invitrogen™
C10407 |
Alexa Fluor™ 594 | DIBO Alkyne | ∼1200 |
£508.00
Each |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
10598370
|
Invitrogen™
C10413 |
Maleimide | DIBO Alkyne, Maleimide | ∼440 | N/A | ||||
15848836
|
Invitrogen™
C20020 |
Alexa Fluor™ 488 | sDIBO Alkyne | ∼1400 |
£564.00
0.50mg |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
15930573
|
Invitrogen™
C20021 |
Alexa Fluor™ 555 | sDIBO Alkyne | ∼1800 |
£576.00
0.50mg |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
15940573
|
Invitrogen™
C20022 |
Alexa Fluor™ 647 | sDIBO Alkyne | ∼1900 |
£554.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15950573
|
Invitrogen™
C20023 |
Biotin | sDIBO Alkyne | ∼1000 |
£536.00
1mg |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
15970573
|
Invitrogen™
C20025 |
SDP ester | SDP ester, sDIBO Alkyne | ∼1050 |
£481.00
1mg |
Estimated Shipment: 09-12-2024
Log in to see stock. |
|||
Description
Achieve efficient labeling of azides with Click-It sDIBO Alkynes. Our Click-IT sDIBO alkynes enable the labeling of azido labeled targets without the need for copper catalysis and are available with a variety of Alexa Fluor or biotin labels, as well as amine or thiol reactive groups for creating your own conjugates. Macromolecules that have been azido-modified enzymatically, chemically, or metabolically can now be labeled easily, yielding soluble bioconjugates.
Click-iT sDIBO Alkyne labels react with azides via a copper-free Click chemistry reaction to form a stable triazole linkage with an Alexa Fluor, Biotin, or label of your choice. Copper-free click reactions using our sDIBO alkynes are ideal for surface labeling of live cells; they also minimize damage to fluorescent proteins like GFP or R-PE. For intracellular labeling, the nonspecific binding of DIBO alkynes creates a background signal, and a labeling strategy using picolyl azides is the preferred solution in these instances.
Click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background. Macromolecules that have been azide-modified enzymatically, chemically, or metabolically can be labeled more easily, yielding more soluble bioconjugates with improved biological labeling utility.
To make a concentrated stock solution from 2–5 mM, dissolve the dye-alkyne in high quality anhydrous DMSO. Store frozen, desiccated, and protected from light. Click-iT SDP Ester sDIBO alkyne is susceptible to hydrolysis, so prepare the stock solution fresh on the day of the experiment.
Specifications
Less hazardous | |
Click Chemistry | |
Dry Ice | |
Azide |