missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Clifton™ aluminum Boiling Bath
Click to view available options
Model:
1105 E
1105 ER
Controller Type:
Constant boiling, no control
Gentle simmer to vigorous boil
Temperature Range:
Constant boiling at 100°C
Simmer to boiling at 100°C
Unit Size:
Each
Description
Rectangular, aluminum electrically heated, multi-place baths regulate from a gentle simmer to a vigorous boil, fitted with concentric ringed lid to vary the hole size.
The steam produced provides an effective source of heat for samples:
The steam produced provides an effective source of heat for samples:
- that may be otherwise thermally damaged by a naked flame.
- used for applications to distill/concentrate liquids, melt agar and other solids.
- metallurgical analysis, protein analysis and bacteriological examinations
- Features an integral constant level device to maintain constant water level during boiling and supplied with a set of rings to vary the diameter of aperture to accommodate different sizes of vessels of evaporating dishes and glassware, including round-bottom flasks.
- Fitted with a sheathed immersion element and a one shot thermal cut out for safety.
- Warranty: 2 year
Specifications
Specifications
| Controller Type | Constant boiling, no control |
| Description | Boiling Bath |
| Capacity (Metric) | 3.12 L |
| Temperature Range | Constant boiling at 100°C |
| For Use With (Application) | Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana |
| Material | Aluminum |
| Model | 1105 E |
| Shape | Rectangular |
| Warranty | 2 Years |
| Dimensions (D x W x H) | 200 x 400 x 90 mm |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction