missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Clifton™ Stainless Steel Basket

Complete an Ultrasonic Bath with a stainless steel basket. Nickel Electro™ Clifton™ Stainless Steel Basket is durable and made of high quality stainless steel.

Brand:  Nickel Electro™ BSK-12

 View more versions of this product

Product Code. 13121323

  • £318.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High quality
  • Durable
  • Stainless steel
  • For use with Ultrasonic Bath
  • Includes side lifting handles
Specifications

Specifications

1 Ea.
12 L
Ultrasonic Bath, 12 L
115 mm
250 x 190 x 115 mm
Basket
190 mm
For Ultrasonic Bath
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.