missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Stainless Steel Rectangular Electric Boiling Baths
Get optimum boiling results with this rectangular, stainless steel electrically heated boiling bath. Featuring an energy regulator that controls the amount of boiling, this Clifton™ Stainless Steel Rectangular Electric Boiling Bath is ideal for safety.
£760.00 - £1105.00
Specifications
Description | Boiling Bath |
---|---|
For Use With (Application) | Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana |
Material | Stainless Steel |
Shape | Rectangular |
Warranty | 2 Years |
Product Code | Brand | Model | Depth (Metric) | Capacity (Metric) | Temperature Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Depth (Metric) | Capacity (Metric) | Temperature Range | Price | Quantity & Availability | ||
12718878
|
Nickel Electro™
1104E |
1104 E | 321 mm | 8.8 L | Constant boiling at 100°C |
£1068.12
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
12799326
|
Nickel Electro™
1102E |
1102 E | 321 mm | 8.8 L | Constant boiling at 100°C |
£760.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||
12798868
|
Nickel Electro™
1104ER |
1104 ER | 321 mm | 8.8 L | Simmer to boiling at 100°C |
£1105.00
Each |
In Stock
Log in to see stock. |
||
12788868
|
Nickel Electro™
1101ER |
1101 ER | 219 mm | 4.54 L | Simmer to boiling at 100°C |
£931.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||
Description
- Rectangular, stainless steel electrically heated
- Multi-place baths with fixed temperature for boiling water
- Fitted with concentric ringed lids to vary the hole size
- Features an integral constant level device to maintain constant water level during boiling
- Fitted with a sheathed immersion element and a one shot thermal cut out for safety
Specifications
Boiling Bath | |
Stainless Steel | |
2 Years | |
230 V |
Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana | |
Rectangular | |
90 mm |