missing translation for 'onlineSavingsMsg'
Learn More

Clindamycin, White Powder, MP Biomedicals™

Product Code. 11466171
Click to view available options
Quantity:
10 mg
50 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 21462-39-5
Molecular Formula C18H34Cl2N2O5S
Molecular Weight (g/mol) 461.439
MDL Number MFCD07793327
InChI Key AUODDLQVRAJAJM-PIPVPTKYSA-N
Synonym (7S)-7-Chloro-7-deoxylincomycin hydrochloride, Cleocin
PubChem CID 134129519
IUPAC Name (2S,4R)-N-[(1S,2S)-2-chloro-1-[(2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-methylsulfanyloxan-2-yl]propyl]-1-methyl-4-propylpyrrolidine-2-carboxamide;hydrochloride
SMILES CCCC1CC(N(C1)C)C(=O)NC(C2C(C(C(C(O2)SC)O)O)O)C(C)Cl.Cl

Specifications

Type Clindamycin
Physical Form Solid
Formula Weight 461.4
Color White
Quantity 10 mg
Chemical Name or Material Clindamycin
compliance-icons
Product Identifier
  • Clindamycin Hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : clindamycim hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'