Learn More
Jenway™ PFP7/C Clinical Flame Photometer
Designed for use in clinical applications for the determination of sodium, potassium and lithium. Jenway™ PFP7/C Clinical Flame Photometer features built-in linearizer circuitry that allows readings of both sodium and potassium to be displayed directly in mmol/l.
Brand:  Jenway™ 12655-00
Product Code. 11495059
  • ONLINE EXCLUSIVE PRICE
    £5355.00
    £4900.00 / Each
    Save £455.00
    8% Off
Only null left
Estimated Shipment: 07-05-2026
Add to basket
Description
Description
  • Designed for clinical analysis
  • Supplied with Na, K, Li, Ba and Ca filters
  • Low temperature, single channel
  • Flame failure safety system
  • Operates with propane, butane, natural gas or LPG
  • 3 year warranty
Specifications
Specifications
Clinical Flame Photometer PFP7